Learn More
Azzota Corp 10mm Pathlength Fluorometer Cuvettes - 0.70Ml Four (4) Sides Clear Pathlength - 10mm Inside Width - 2mm Volume - 0.70Ml Dimension - 45X12.5X12.5mm With Cover And Round Bottom Quartz (190Nm-2500Nm)
Catalog No. 507532896
Encompass_Preferred
10mm Pathlength Fluorometer Cuvettes - 0.70ml Four (4) sides clear Pathlength - 10mm Inside width - 2mm Volume - 0.70ml Dimension - 45x12.5x12.5mm With cover and round bottom Quartz (190nm-2500nm)
Catalog No. 50-753-2896 Supplier Azzota Corp Supplier No. Q514
10mm Pathlength Fluorometer Cuvettes - 0.70ml Four (4) sides clear Pathlength - 10mm Inside width - 2mm Volume - 0.70ml Dimension - 45x12.5x12.5mm With cover and round bottom Quartz (190nm-2500nm)