Learn More
BIOSMITH Consumables. 2mm gap electroporation cuvette. Fits all commercial electroporators. Single packaged. 50 per pack
Catalog No. NC2980201
Encompass
Consumables. 2mm gap electroporation cuvette. Fits all commercial electroporators. Single packaged. 50 per pack
Catalog No. NC2980201 Supplier BIOSMITH Supplier No. 72002
Consumables. 2mm gap electroporation cuvette. Fits all commercial electroporators. Single packaged. 50 per pack