Learn More
Chemglass Life Sciences 2mm Bore, 12/30 Plug Size, 8mm OD Sidearm, #1 Clip Size
SDP
Catalog No. 501531989
Encompass_Preferred
2mm Bore, 12/30 Plug Size, 8mm OD Sidearm, #1 Clip Size. Straight bore glass Stopcocks are supplied complete with the listed size metal retaining clip CG-423.
Catalog No. 50-153-1989 Supplier Chemglass Life Sciences Supplier No. CGB40002
2mm Bore, 12/30 Plug Size, 8mm OD Sidearm, #1 Clip Size. Straight bore glass Stopcocks are supplied complete with the listed size metal retaining clip CG-423.