Learn More
Chemglass Life Sciences Stopcock, Straight Bore, Glass Plug, High Vacuum, 2mm Bore, 12/30 Plug Size, 8mm OD Sidearms
SDP
Catalog No. CG46201
Encompass_Preferred
Solid glass high vacuum stopcock with theChemglass improved constant pressure retention device. This retaining system incorporates a glass plug having an extended snelling which accepts a stainless steel spring and clip. The spring keeps constant tension on the plug at all times and is easily removed and attached. Plug is precision ground and hand-lapped with corresponding numbers on the barrel and plug. Each stopcock is supplied complete with a stainless steel spring and clip. Replacement retention kit consists of one spring and clip for the plug size listed.
Catalog No. CG46201 Supplier Chemglass Life Sciences Supplier No. CG46201
Solid glass high vacuum stopcock with theChemglass improved constant pressure retention device. This retaining system incorporates a glass plug having an extended snelling which accepts a stainless steel spring and clip. The spring keeps constant tension on the plug at all times and is easily removed and attached. Plug is precision ground and hand-lapped with corresponding numbers on the barrel and plug. Each stopcock is supplied complete with a stainless steel spring and clip. Replacement retention kit consists of one spring and clip for the plug size listed.