Learn More
Chemglass Life Sciences 2mm Bore, 11/25 Plug Size, 8mm OD Sidearm
SDP
Catalog No. 501531991
Encompass_Preferred
2mm Bore, 11/25 Plug Size, 8mm OD Sidearm. Straight bore stopcock having a 1:5 taper solid PTFE plug.
Catalog No. 50-153-1991 Supplier Chemglass Life Sciences Supplier No. CGB42403
2mm Bore, 11/25 Plug Size, 8mm OD Sidearm. Straight bore stopcock having a 1:5 taper solid PTFE plug.