Learn More
Chemglass Life Sciences Stopcock, Straight Bore, Both Arms Capillary, PTFE Plug, 2mm Bore, 11/25 Plug Size, 7mm OD Sidearms
SDP
Catalog No. CG43201
Encompass_Preferred
Stopcock, Straight Bore, Both Arms Capillary, PTFE Plug, 2mm Bore, 11/25 Plug Size, 7mm OD Sidearms. Similar to CG-424, but with both arms being made of 2mm ID capillary tubing.
Catalog No. CG43201 Supplier Chemglass Life Sciences Supplier No. CG43201
Stopcock, Straight Bore, Both Arms Capillary, PTFE Plug, 2mm Bore, 11/25 Plug Size, 7mm OD Sidearms. Similar to CG-424, but with both arms being made of 2mm ID capillary tubing.