Learn More
Chemglass Life Sciences Stopcock, Straight Bore, 1-Arm Capillary, PTFE Plug, 2mm Bore, 11/25 Plug Size, 7mm & 8mm OD Sidearms
SDP
Catalog No. CG43001
Encompass_Preferred
Stopcock, Straight Bore, 1-Arm Capillary, PTFE Plug, 2mm Bore, 11/25 Plug Size, 7mm & 8mm OD Sidearms. Similar to CG-424, but with one arm being made of 2mm ID capillary tubing.
Catalog No. CG43001 Supplier Chemglass Life Sciences Supplier No. CG43001
Stopcock, Straight Bore, 1-Arm Capillary, PTFE Plug, 2mm Bore, 11/25 Plug Size, 7mm & 8mm OD Sidearms. Similar to CG-424, but with one arm being made of 2mm ID capillary tubing.