Learn More
Elemental Scientific Inc 2MM QTZ INJCT X SERIES/1305760
SDP
Catalog No. NC0376007
Encompass_Preferred
2mm i.d. quartz o-ring-free injector for Thermo XSeries. Use with Precision quartz torch and ESI spray chamber.
Catalog No. NC0376007 Supplier Elemental Scientific Inc Supplier No. ES12333200
2mm i.d. quartz o-ring-free injector for Thermo XSeries. Use with Precision quartz torch and ESI spray chamber.