Learn More
Azzota Corp 10mm Pathlength Microcuvettes 0.7Ml - Pathlength 10mm - Inside Width 2mm Volume 0.7Ml Dimensions 45X12.5X12.5mm; Cover And Black Walls, Glass (340Nm-2500Nm)
Catalog No. 507532905
Encompass_Preferred
10mm Pathlength MicroCuvettes 0.7ml - Pathlength 10mm - Inside width 2mm Volume 0.7ml Dimensions 45x12.5x12.5mm; Cover and black walls, Glass (340nm-2500nm)
Catalog No. 50-753-2905 Supplier Azzota Corp Supplier No. Q44
10mm Pathlength MicroCuvettes 0.7ml - Pathlength 10mm - Inside width 2mm Volume 0.7ml Dimensions 45x12.5x12.5mm; Cover and black walls, Glass (340nm-2500nm)