Learn More
Alkali Scientific 2mm Electroporation Cuvettes, Square Lid, Sterile, 50/cs
SDP
Catalog No. 502335890
Encompass_Preferred
Manufactured from high quality durable polycarbonate material which has been tested to optimize transfection efficiencies. Precision built to give exacting tolerances ensuring reproducible results. Polished aluminum plates reduce static and arcing for optimum performance. Individually wrapped and gamma irradiated providing guaranteed sterility. Compatible with most Electroporation devices. Individually color coded cuvette caps identify gap widths of 2mm. Each bag contains 50 individually wrapped sterile cuvettes.
Catalog No. 50-233-5890 Supplier Alkali Scientific Supplier No. EP402002
Manufactured from high quality durable polycarbonate material which has been tested to optimize transfection efficiencies. Precision built to give exacting tolerances ensuring reproducible results. Polished aluminum plates reduce static and arcing for optimum performance. Individually wrapped and gamma irradiated providing guaranteed sterility. Compatible with most Electroporation devices. Individually color coded cuvette caps identify gap widths of 2mm. Each bag contains 50 individually wrapped sterile cuvettes.