Learn More
Ace Glass, Inc. 2MM STOPCOCK (GB)
SDP
Catalog No. 50976136
Encompass_Preferred
2mm Newman type stopcock is used with a distilling head to enable the operator to determine reflux ratios and take-offs. Process is described in Industrial and Engineering Chemistry, Volume 14, No. 11, Page 902, Nov. 15, 1942.
Catalog No. 50-976-136 Supplier Ace Glass, Inc. Supplier No. 815204
2mm Newman type stopcock is used with a distilling head to enable the operator to determine reflux ratios and take-offs. Process is described in Industrial and Engineering Chemistry, Volume 14, No. 11, Page 902, Nov. 15, 1942.