Learn More
Chemglass Life Sciences Stopcock, Straight Bore, Glass Plug, Capillary Arms, 2mm Bore, 12/30 Plug Size, 7.5mm OD Sidearms, #1 Clip Size
SDP
Catalog No. CG40602
Encompass_Preferred
Stopcock, Straight Bore, Glass Plug, Capillary Arms, 2mm Bore, 12/30 Plug Size, 7.5mm OD Sidearms, #1 Clip Size. Similar to CG-400, but with the sidearms being made of capillary tubing having the same bore as the plug. Supplied complete with metal retaining clip CG-423.*** FOR REPLACEMENT PLUGS, SEE CG-400 LISTING ***
Catalog No. CG40602 Supplier Chemglass Life Sciences Supplier No. CG40602
Stopcock, Straight Bore, Glass Plug, Capillary Arms, 2mm Bore, 12/30 Plug Size, 7.5mm OD Sidearms, #1 Clip Size. Similar to CG-400, but with the sidearms being made of capillary tubing having the same bore as the plug. Supplied complete with metal retaining clip CG-423.*** FOR REPLACEMENT PLUGS, SEE CG-400 LISTING ***