Learn More
Chemglass Life Sciences Stopcock, Straight Bore, Glass Plug, Schlenk, 2mm Bore, 12/30 Plug Size, Plain Snelling
SDP
Catalog No. 50873713
Encompass_Preferred
Stopcock, Straight Bore, Glass Plug, Schlenk, 2mm Bore, 12/30 Plug Size, Plain Snelling. This stopcock is especially designed for the repair of the sidearm on Schlenk vessels. Stopcocks are available with a standard snelling to accept a CG-423-01 metal retaining clip. Stopcock has a heavy duty, larger diameter (10mm OD) sidearm which makes an exceptionally strong seal to the Schlenk vessel. The hose connection has an 8mm OD and is 35mm in length. For replacement components see CG-400.
Catalog No. 50-873-713 Supplier Chemglass Life Sciences Supplier No. CG40901
Stopcock, Straight Bore, Glass Plug, Schlenk, 2mm Bore, 12/30 Plug Size, Plain Snelling. This stopcock is especially designed for the repair of the sidearm on Schlenk vessels. Stopcocks are available with a standard snelling to accept a CG-423-01 metal retaining clip. Stopcock has a heavy duty, larger diameter (10mm OD) sidearm which makes an exceptionally strong seal to the Schlenk vessel. The hose connection has an 8mm OD and is 35mm in length. For replacement components see CG-400.