Learn More
Sutter Instruments 2.0 mm Wide Trough Filament
SDP
Catalog No. NC0442567
Encompass_Preferred
Excellent general purpose filaments and are recommended for creating longer slice patch pipettes and sharp electrodes. The trough filament is not ideal for making patch pipettes or when using glass over 1.2 mm OD. If very short tapers are required for patch electrodes or for microinjection, a 2.5 mm or 3 mm box filament is recommended.
Catalog No. NC0442567 Supplier Sutter Instruments Supplier No. FGFT320B
Excellent general purpose filaments and are recommended for creating longer slice patch pipettes and sharp electrodes. The trough filament is not ideal for making patch pipettes or when using glass over 1.2 mm OD. If very short tapers are required for patch electrodes or for microinjection, a 2.5 mm or 3 mm box filament is recommended.