Learn More
Cayman Chemical 2mthylaPyrroldInobutIoph 10mg
SDP
Catalog No. 502892824
Encompass_Preferred
An isomer of 4-methyl PBP, having the methyl group attached at the 2 position of the phenyl ring, instead of the 4 position; intended for forensic and research applications
Catalog No. 50-289-2824 Supplier Cayman Chemical Supplier No. 900118810 MG
An isomer of 4-methyl PBP, having the methyl group attached at the 2 position of the phenyl ring, instead of the 4 position; intended for forensic and research applications