Learn More
Cayman Chemical 2mthylaPyrroldInobutIoph 50mg
SDP
Catalog No. 502896099
Encompass_Preferred
An isomer of 4-methyl PBP, having the methyl group attached at the 2 position of the phenyl ring, instead of the 4 position; intended for forensic and research applications
Catalog No. 50-289-6099 Supplier Cayman Chemical Supplier No. 900118850 MG
An isomer of 4-methyl PBP, having the methyl group attached at the 2 position of the phenyl ring, instead of the 4 position; intended for forensic and research applications