Learn More
Cayman Chemical 2-methyl-α-Pyrrolidinopropiophenone (hydrochloride), Purity: ≥98%, CAS: 2749897-10-5, 50 mg
SDP
Catalog No. 502951127
Encompass_Preferred
Shares structural features with the stimulant α-PPP and is a positional isomer of 4'-methyl-α-PPP (hydrochloride), which has been detected in bath salts and other formulations; this product is intended to be used for forensic and research applications
Catalog No. 50-295-1127 Supplier Cayman Chemical Supplier No. 1148450 MG
Shares structural features with the stimulant α-PPP and is a positional isomer of 4'-methyl-α-PPP (hydrochloride), which has been detected in bath salts and other formulations; this product is intended to be used for forensic and research applications