Learn More
Cayman Chemical 2mthylthIoadenosIn dIphsp 10mg
SDP
Catalog No. 502985547
Encompass_Preferred
An agonist of P2Y receptors (EC50s = 5.13, 0.89, 0.58, 0.89, and 6.2 for human P2Y1 and P2Y12, rat P2Y1 and P2Y12, and mouse P2Y13 receptors, respectively); induces platelet aggregation and inhibits cyclic AMP accumulation in PGE1-stimulated platelet-rich plasma more potently than ADP
Catalog No. 50-298-5547 Supplier Cayman Chemical Supplier No. 2123010 MG
An agonist of P2Y receptors (EC50s = 5.13, 0.89, 0.58, 0.89, and 6.2 for human P2Y1 and P2Y12, rat P2Y1 and P2Y12, and mouse P2Y13 receptors, respectively); induces platelet aggregation and inhibits cyclic AMP accumulation in PGE1-stimulated platelet-rich plasma more potently than ADP