Learn More
Cayman Chemical 2-Methylthioadenosine triphosphate (sodium salt), Purity: ≥95%, CAS: 100020-57-3, 2.8 mg
SDP
Catalog No. 502994029
Encompass_Preferred
A P2X2 and P2Y1 receptor agonist; inhibits GC-C in rat intestinal mucosal membranes (Ki = 360 µM in the presence of E. coli heat-stable enterotoxin); induces calcium influx in primary rat arcuate nucleus neurons in a fluorescence-based reporter assay at 1 µM; increases inositol phosphate accumulation in 1321N1 astrocytoma cells expressing human P2Y1 receptors (EC50 = 51 nM); inhibits uroguanylin-induced increases in urine flow and sodium excretion in rats
Catalog No. 50-299-4029 Supplier Cayman Chemical Supplier No. 214182.8 MG
A P2X2 and P2Y1 receptor agonist; inhibits GC-C in rat intestinal mucosal membranes (Ki = 360 µM in the presence of E. coli heat-stable enterotoxin); induces calcium influx in primary rat arcuate nucleus neurons in a fluorescence-based reporter assay at 1 µM; increases inositol phosphate accumulation in 1321N1 astrocytoma cells expressing human P2Y1 receptors (EC50 = 51 nM); inhibits uroguanylin-induced increases in urine flow and sodium excretion in rats