Learn More
Chemglass Life Sciences 2000mL 2-Neck Round Bottom Flask (RBF) , 1-CN 24/40 Outer, 1-SN 24/40 Outer
SDP
Catalog No. CG151810
Encompass_Preferred
2000mL 2-Neck Round Bottomed Flask, 1-CN 24/40 Outer, 1-SN 24/40 Outer. Round bottom, HEAVY WALL two neck flask with standard taper outer joints on both the center neck and vertical side neck.
Catalog No. CG151810 Supplier Chemglass Life Sciences Supplier No. CG151810
2000mL 2-Neck Round Bottomed Flask, 1-CN 24/40 Outer, 1-SN 24/40 Outer. Round bottom, HEAVY WALL two neck flask with standard taper outer joints on both the center neck and vertical side neck.