Learn More
Cayman Chemical 2-nonyl-3-hydroxy-4-Quinolone, Purity: ≥98%, CAS: 1259944-03-0, 10 mg
SDP
Catalog No. 502893647
Encompass_Preferred
A bacterial quorum-sensing signaling molecule; activates a biosensor for the P. aeruginosa gene pqsA in a concentration-dependent manner
Catalog No. 50-289-3647 Supplier Cayman Chemical Supplier No. 900269910 MG
A bacterial quorum-sensing signaling molecule; activates a biosensor for the P. aeruginosa gene pqsA in a concentration-dependent manner