Learn More
Cayman Chemical 2-O-(α-D-Glucopyranosyl)glycerol, Purity: ≥95%, CAS: 22160-26-5, 10 g
SDP
Catalog No. 502946500
Encompass_Preferred
A compatible solute; prevents drying-induced membrane fusion and CF leakage in liposomes; levels are increased in response to osmotic stress in bacteria
Catalog No. 50-294-6500 Supplier Cayman Chemical Supplier No. 3449210 G
A compatible solute; prevents drying-induced membrane fusion and CF leakage in liposomes; levels are increased in response to osmotic stress in bacteria