Learn More
Cayman Chemical 2-O-α-D-Glucopyranosyl-L-ascorbic Acid, Purity: ≥95%, CAS: 129499-78-1, 25 g
SDP
Catalog No. 502991712
Encompass_Preferred
An L-ascorbic acid derivative with antioxidant and radioprotective activities; scavenges DPPH radicals in a cell-free assay (EC50 = 61 μM in 60% ethanol); inhibits γ-radiation-induced cell death in CHO-10B2 and radiosensitive xrs5 cells, as well as UVC- or broadband UVB-induced cell death in CHO-10B2 and UV-sensitive UV135 cells, at 5 mM; increases serum levels of ascorbic acid in rats and guinea pigs at 19.2 and 96 mg/animal, p.o., respectively; reverses weight loss and inhibits subcutaneous hemorrhage in guinea pigs fed a vitamin C-deficient diet at 19.2 mg/animal per day
Catalog No. 50-299-1712 Supplier Cayman Chemical Supplier No. 3140225 G
An L-ascorbic acid derivative with antioxidant and radioprotective activities; scavenges DPPH radicals in a cell-free assay (EC50 = 61 μM in 60% ethanol); inhibits γ-radiation-induced cell death in CHO-10B2 and radiosensitive xrs5 cells, as well as UVC- or broadband UVB-induced cell death in CHO-10B2 and UV-sensitive UV135 cells, at 5 mM; increases serum levels of ascorbic acid in rats and guinea pigs at 19.2 and 96 mg/animal, p.o., respectively; reverses weight loss and inhibits subcutaneous hemorrhage in guinea pigs fed a vitamin C-deficient diet at 19.2 mg/animal per day