Learn More
Selleck Chemical LLC 2OH-BNPP1 S3522-25mg
SDP
Catalog No. 503108960
Encompass_Preferred
2OH-BNPP1 is an inhibitor of the kinase activity of BUB1 (budding uninhibited by benzimidazoles-1 BUB1 Mitotic Checkpoint Serine/Threonine Kinase) that can be used for the treatment of cancer
Catalog No. 50-310-8960 Supplier Selleck Chemical LLC Supplier No. S352225MG
2OH-BNPP1 is an inhibitor of the kinase activity of BUB1 (budding uninhibited by benzimidazoles-1 BUB1 Mitotic Checkpoint Serine/Threonine Kinase) that can be used for the treatment of cancer