Learn More
Selleck Chemical LLC 2OH-BNPP1 S3522-5mg
SDP
Catalog No. 503108736
Encompass_Preferred
2OH-BNPP1 is an inhibitor of the kinase activity of BUB1 (budding uninhibited by benzimidazoles-1 BUB1 Mitotic Checkpoint Serine/Threonine Kinase) that can be used for the treatment of cancer
Catalog No. 50-310-8736 Supplier Selleck Chemical LLC Supplier No. S35225MG
2OH-BNPP1 is an inhibitor of the kinase activity of BUB1 (budding uninhibited by benzimidazoles-1 BUB1 Mitotic Checkpoint Serine/Threonine Kinase) that can be used for the treatment of cancer