Learn More
TARGETMOL CHEMICALS INC 2OH-BNPP1 5MG
SDP
Catalog No. 502880098
Encompass_Preferred
Also available in 1 mL, 1 mg, 2 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. 2OH-BNPP1 is a BUB1 kinase, a Ser/Thr kinase inhibitor, it used for the treatment of cancer. Purity 98.45%
Catalog No. 50-288-0098 Supplier TARGETMOL CHEMICALS INC Supplier No. T140225MG
Also available in 1 mL, 1 mg, 2 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. 2OH-BNPP1 is a BUB1 kinase, a Ser/Thr kinase inhibitor, it used for the treatment of cancer. Purity 98.45%