Learn More
Cayman Chemical 2-O-methyl PAF C-18 (hydrate), Purity: ≥98%, 100 mg
SDP
Catalog No. 502892487
Encompass_Preferred
A synthetic PAF analog which contains an ether-linked methyl group at the sn-2 position; decreases plasma membrane fluidity, inhibits tumor cell invasiveness, and causes the release of large quantities of NO by a pathway involving activation of NOS
Catalog No. 50-289-2487 Supplier Cayman Chemical Supplier No. 60912100 MG
A synthetic PAF analog which contains an ether-linked methyl group at the sn-2 position; decreases plasma membrane fluidity, inhibits tumor cell invasiveness, and causes the release of large quantities of NO by a pathway involving activation of NOS