Learn More
Cayman Chemical 2'-O-Methyl-5-methyluridine, Purity: ≥98%, CAS: 55486-09-4, 250 mg
SDP
Catalog No. 502913399
Encompass_Preferred
A modified nucleoside; has been found in tRNA isolated from rabbit liver
Catalog No. 50-291-3399 Supplier Cayman Chemical Supplier No. 35357250 MG
A modified nucleoside; has been found in tRNA isolated from rabbit liver