Learn More
Cayman Chemical 2OMethylgunosIn5OtrI 2.7mg
SDP
Catalog No. 502918946
Encompass_Preferred
An HCV NS5B inhibitor (IC50 = 3.5 µM) and active metabolite of IDX184; induces tubulin polymerization at 50 µM; tumoral levels are decreased in patients with hepatocellular carcinoma compared with adjacent normal tissue
Catalog No. 50-291-8946 Supplier Cayman Chemical Supplier No. 385682.7 MG
An HCV NS5B inhibitor (IC50 = 3.5 µM) and active metabolite of IDX184; induces tubulin polymerization at 50 µM; tumoral levels are decreased in patients with hepatocellular carcinoma compared with adjacent normal tissue