Learn More
Cayman Chemical 2oxo3hydroxy LSdolutIon 100ug
SDP
Catalog No. 502945511
Encompass_Preferred
An analytical reference standard categorized as a lysergamide; a metabolite of LSD; intended for research and forensic applications
Catalog No. 50-294-5511 Supplier Cayman Chemical Supplier No. 41933100 UG
An analytical reference standard categorized as a lysergamide; a metabolite of LSD; intended for research and forensic applications