Learn More
C&G Containers 2 Oz Clear Glass French Square Bottle, High Chemical Resistance, Efficient Storage, 2 oz
SDP
Catalog No. NC1205350
Encompass_Preferred
Introducing a 2 oz (60 mL) Clear Glass French Square Bottle, designed for effective solvent and chemical storage. Its superior chemical resistance, coupled with a PTFE-faced liner, ensures product integrity while optimizing space with its square shape. Ideal for laboratory applications, this bottle comes with a white polypropylene cap, perfect for secure containment. Elevate your lab's efficiency with this reliable storage solution.
This item is not returnable. View return policy
Catalog No. NC1205350 Supplier C&G Containers Supplier No. LFF002308500
Introducing a 2 oz (60 mL) Clear Glass French Square Bottle, designed for effective solvent and chemical storage. Its superior chemical resistance, coupled with a PTFE-faced liner, ensures product integrity while optimizing space with its square shape. Ideal for laboratory applications, this bottle comes with a white polypropylene cap, perfect for secure containment. Elevate your lab's efficiency with this reliable storage solution.