Learn More
Cayman Chemical 2Phenyl2 1pIperIdInylpro 10mg
SDP
Catalog No. 502966499
Encompass_Preferred
A mechanism-based inactivator of human CYP2B6 (Ki = 5.6 µM; IC50 = 5.1 µM) with potent selectivity for CYP2B6 over other CYP450 isoforms
Catalog No. 50-296-6499 Supplier Cayman Chemical Supplier No. 1779910 MG
A mechanism-based inactivator of human CYP2B6 (Ki = 5.6 µM; IC50 = 5.1 µM) with potent selectivity for CYP2B6 over other CYP450 isoforms