Learn More
Cayman Chemical 2propyl4PentenoIc AcId 100mg
SDP
Catalog No. 502993896
Encompass_Preferred
A metabolite of valproic acid; undergoes β-oxidation to form reactive metabolites that induce hepatotoxicity in rats; neuroteratogenic, reducing forebrain size and inducing deformation of the neural tube in mouse embryos
Catalog No. 50-299-3896 Supplier Cayman Chemical Supplier No. 23090100 MG
A metabolite of valproic acid; undergoes β-oxidation to form reactive metabolites that induce hepatotoxicity in rats; neuroteratogenic, reducing forebrain size and inducing deformation of the neural tube in mouse embryos