Learn More
Cayman Chemical 2propyl4PentenoIc AcId 10mg
SDP
Catalog No. 502994696
Encompass_Preferred
A metabolite of valproic acid; undergoes β-oxidation to form reactive metabolites that induce hepatotoxicity in rats; neuroteratogenic, reducing forebrain size and inducing deformation of the neural tube in mouse embryos
Catalog No. 50-299-4696 Supplier Cayman Chemical Supplier No. 2309010 MG
A metabolite of valproic acid; undergoes β-oxidation to form reactive metabolites that induce hepatotoxicity in rats; neuroteratogenic, reducing forebrain size and inducing deformation of the neural tube in mouse embryos