Learn More
Cayman Chemical (±)-2-propyl-4-Pentenoic Acid, Purity: ≥98%, CAS: 1575-72-0, 50 mg
SDP
Catalog No. 502994697
Encompass_Preferred
A metabolite of valproic acid; undergoes β-oxidation to form reactive metabolites that induce hepatotoxicity in rats; neuroteratogenic, reducing forebrain size and inducing deformation of the neural tube in mouse embryos
Catalog No. 50-299-4697 Supplier Cayman Chemical Supplier No. 2309050 MG
A metabolite of valproic acid; undergoes β-oxidation to form reactive metabolites that induce hepatotoxicity in rats; neuroteratogenic, reducing forebrain size and inducing deformation of the neural tube in mouse embryos