Learn More
Cayman Chemical 2-p-Toluidinylnaphthylene-6-sulfonate (sodium salt), Purity: ≥98%, CAS: 53313-85-2, 50 mg
SDP
Catalog No. 502915315
Encompass_Preferred
An anionic fluorescent probe for the conformational state of proteins; ex/em = 321/446 nm, respectively; has been used to determine the pKa values of LNPs
Catalog No. 50-291-5315 Supplier Cayman Chemical Supplier No. 3714050 MG
An anionic fluorescent probe for the conformational state of proteins; ex/em = 321/446 nm, respectively; has been used to determine the pKa values of LNPs