Learn More
World Precision Instrument Microinjection Syringe Pump with SMARTouch Controller-UMP3T-2
SDP
Catalog No. NC1987991
Encompass
The touch-screen display on WPI's MICRO2T SMARTouch™ controller offers an intuitive, user-friendly interface. Our MICRO2T allows for simultaneous control of the UMP3 syringe pumps, and/or our NANOLITER2020 positive displacement pumps. Key features of the MICRO2T controller include a splash-proof touch screen, built-in memory configured for saving settings between use, and a dual mode motor drive. The UMP3 may be mounted to either a stereotaxic frame or a micro manipulator via its built-in mounting bar. An optional foot switch is available. 
To learn more about our warranty options, click here.
Catalog No. NC1987991 Supplier World Precision Instrument Supplier No. UMP3T2
Add to Cart
Add to Cart
Request a Quote
The touch-screen display on WPI's MICRO2T SMARTouch™ controller offers an intuitive, user-friendly interface. Our MICRO2T allows for simultaneous control of the UMP3 syringe pumps, and/or our NANOLITER2020 positive displacement pumps. Key features of the MICRO2T controller include a splash-proof touch screen, built-in memory configured for saving settings between use, and a dual mode motor drive. The UMP3 may be mounted to either a stereotaxic frame or a micro manipulator via its built-in mounting bar. An optional foot switch is available. 
To learn more about our warranty options, click here.