Learn More
Medchemexpress LLC (2R)-1,1,1-trifluoro-2-(3-(piperazin-1-ylsulfonyl)phenyl)propan-2-ol | 946396-92-5 | 100.0% | 100 MG
SDP
Catalog No. 5000230555
Encompass_Preferred
HSD-016 is a potent, selective, orally active inhibitor of 11β-hydroxysteroid dehydrogenase type 1 (11β-HSD1), provided as a high-purity research reagent in powder and DMSO solution formats for preclinical biochemical studies.
  • High purity suitable for research applications.
  • Available in multiple solid masses and as ready-to-use DMSO solution.
  • Soluble in DMSO for convenient reconstitution.
  • Suitable for enzymatic inhibition and metabolic pathway studies.
  • Supplied with supporting analytical data where available.
Catalog No. 50-002-30555 Supplier Medchemexpress LLC Supplier No. HY122028100MG
HSD-016 is a potent, selective, orally active inhibitor of 11β-hydroxysteroid dehydrogenase type 1 (11β-HSD1), provided as a high-purity research reagent in powder and DMSO solution formats for preclinical biochemical studies.
  • High purity suitable for research applications.
  • Available in multiple solid masses and as ready-to-use DMSO solution.
  • Soluble in DMSO for convenient reconstitution.
  • Suitable for enzymatic inhibition and metabolic pathway studies.
  • Supplied with supporting analytical data where available.