Learn More
Medchemexpress LLC (2R)-1,1,1-trifluoro-2-[4-[(2S)-2-[[(3S)-3-methylmorpholin-4-yl]methyl]-4-thiophen-2-ylsulfonyl | 1361217-07-3 | >98.0% | 533.63 g·mol⁻1 | C23H30F3N3O4S2 | 50MG
SDP
Catalog No. 5000239276
Encompass_Preferred
AMG-1694 is a potent small-molecule disruptor of the glucokinase-glucokinase regulatory protein (GK-GKRP) interaction. It promotes dissociation of the GK-GKRP complex and has been reported to normalize blood glucose in preclinical rodent models. Supplied as a powder, it is intended for research use in studies of glucokinase regulation and glucose metabolism.
  • Disrupts GK-GKRP interaction to enhance glucokinase activity.
  • Reported to normalize blood glucose in preclinical rodent models.
  • Potent activity with reported low-nanomolar potency.
  • Chemical identifiers: CAS 1361217-07-3; formula C23H30F3N3O4S2; molecular weight 533.63 g·mol⁻1.
  • Purity greater than 98.0% as listed in the supplier catalog.
  • Supplied as a powder with recommended cold storage for stability.
Catalog No. 50-002-39276 Supplier Medchemexpress LLC Supplier No. HY1261450MG
AMG-1694 is a potent small-molecule disruptor of the glucokinase-glucokinase regulatory protein (GK-GKRP) interaction. It promotes dissociation of the GK-GKRP complex and has been reported to normalize blood glucose in preclinical rodent models. Supplied as a powder, it is intended for research use in studies of glucokinase regulation and glucose metabolism.
  • Disrupts GK-GKRP interaction to enhance glucokinase activity.
  • Reported to normalize blood glucose in preclinical rodent models.
  • Potent activity with reported low-nanomolar potency.
  • Chemical identifiers: CAS 1361217-07-3; formula C23H30F3N3O4S2; molecular weight 533.63 g·mol⁻1.
  • Purity greater than 98.0% as listed in the supplier catalog.
  • Supplied as a powder with recommended cold storage for stability.