Learn More
Medchemexpress LLC (2R)-1-((5-(5-(((2,6-dichlorophenyl)methyl-d)thio)-1,3,4-thiadiazol-2-yl)pyrazinyl)amino)propan-2-ol | 3058199-58-6 | 99.8% | 429.37 g/mol | C16H14DCl2N5OS2 | 10 MG
SDP
Catalog No. 5000289307
Encompass_Preferred
A deuterated Piezo1 agonist supplied for laboratory research use. It activates Piezo1 channels and is provided with analytical and safety documentation to support experimental reproducibility and handling.
  • Deuterated Piezo1 agonist suitable for research applications.
  • High purity (99.8%) for experimental consistency.
  • Supplied with datasheet, certificate of analysis, HNMR, and RP-HPLC documentation.
  • Available in multiple small pack sizes for dose-ranging studies.
  • Powder stable at -20°C for up to 3 years; aliquots in solvent require -80°C for long-term storage.
Catalog No. 50-002-89307 Supplier Medchemexpress LLC Supplier No. HY165607S10MG
A deuterated Piezo1 agonist supplied for laboratory research use. It activates Piezo1 channels and is provided with analytical and safety documentation to support experimental reproducibility and handling.
  • Deuterated Piezo1 agonist suitable for research applications.
  • High purity (99.8%) for experimental consistency.
  • Supplied with datasheet, certificate of analysis, HNMR, and RP-HPLC documentation.
  • Available in multiple small pack sizes for dose-ranging studies.
  • Powder stable at -20°C for up to 3 years; aliquots in solvent require -80°C for long-term storage.