Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC [(2R)-2,3-di(hexadecanoyloxy)propyl] 2-(methylazaniumyl)ethyl phosphate | 3930-13-0 | 98.0% | 705.99 g/mol | C38H76NO8P | 1 MG
SDP

Catalog No. 5000284667
Encompass_Preferred

16:0 Monomethyl PE (1,2-NMeDPPE) is a synthetic phosphatidylethanolamine derivative supplied for research use, commonly used in studies of lipid membranes and model membrane systems.

  • Used in lipid and membrane research
  • High purity: 98.0%
  • Chemical formula: C38H76NO8P
  • Molecular weight: 705.99 g/mol
  • CAS number: 3930-13-0
  • Supplied in small milligram quantities suitable for analytical and preparative studies

Catalog No. 50-002-84667 Supplier Medchemexpress LLC Supplier No. HYW7031021MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

16:0 Monomethyl PE (1,2-NMeDPPE) is a synthetic phosphatidylethanolamine derivative supplied for research use, commonly used in studies of lipid membranes and model membrane systems.

  • Used in lipid and membrane research
  • High purity: 98.0%
  • Chemical formula: C38H76NO8P
  • Molecular weight: 705.99 g/mol
  • CAS number: 3930-13-0
  • Supplied in small milligram quantities suitable for analytical and preparative studies

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.