Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (2R)-2-hydroxy-2-methyl-4-(2,4,5-trimethyl-3,6-dioxocyclohexa-1,4-dien-1-yl)butanamide | 1147883-03-1 | 99.3% | 265.30 g/mol | C14H19NO4 | 25MG
SDP

Catalog No. 5000236545
Encompass_Preferred

EPI-589 is a redox-active small-molecule neuroprotective compound used in research on oxidative stress and neurodegenerative disorders. Supplied as a light yellow powder, it is formulated for both in vitro and in vivo experimentation and is readily soluble in DMSO with recommended vehicle formulations for animal dosing. The compound is characterized and shelf-stable under appropriate storage conditions.

  • High purity: 99.3%.
  • Chemical formula: C14H19NO4.
  • Molecular weight: 265.30 g/mol.
  • Form: solid powder, light yellow to yellow.
  • In vitro solubility: DMSO 100 mg/mL.
  • Recommended in vivo vehicles and clear solution ≥ 2.5 mg/mL.
  • Storage: powder at -20°C (long term) or 4°C (short term).

Catalog No. 50-002-36545 Supplier Medchemexpress LLC Supplier No. HY12599925MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

EPI-589 is a redox-active small-molecule neuroprotective compound used in research on oxidative stress and neurodegenerative disorders. Supplied as a light yellow powder, it is formulated for both in vitro and in vivo experimentation and is readily soluble in DMSO with recommended vehicle formulations for animal dosing. The compound is characterized and shelf-stable under appropriate storage conditions.

  • High purity: 99.3%.
  • Chemical formula: C14H19NO4.
  • Molecular weight: 265.30 g/mol.
  • Form: solid powder, light yellow to yellow.
  • In vitro solubility: DMSO 100 mg/mL.
  • Recommended in vivo vehicles and clear solution ≥ 2.5 mg/mL.
  • Storage: powder at -20°C (long term) or 4°C (short term).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.