Learn More
Medchemexpress LLC (2R)-2-hydroxy-2-methyl-4-(2,4,5-trimethyl-3,6-dioxocyclohexa-1,4-dien-1-yl)butanamide | 1147883-03-1 | 99.3% | 265.30 g/mol | C14H19NO4 | 5MG
SDP
Catalog No. 5000235007
Encompass_Preferred
EPI-589 is a redox-active quinone research compound that acts as an oxidoreductase inhibitor and free-radical scavenger. It has shown neuroprotective effects in preclinical models and is used for mechanistic studies and pharmacology experiments. The compound is supplied as a powder for laboratory research and requires appropriate solvent formulation for biological testing.
  • Redox-active oxidoreductase inhibitor.
  • Demonstrated neuroprotective activity in preclinical models.
  • Suitable for in vitro and in vivo pharmacology studies.
  • High reported purity (approximately 99.3%).
  • Soluble in DMSO; established in vivo formulation options reported.
  • Powder stable under low-temperature storage conditions.
Catalog No. 50-002-35007 Supplier Medchemexpress LLC Supplier No. HY1259995MG
EPI-589 is a redox-active quinone research compound that acts as an oxidoreductase inhibitor and free-radical scavenger. It has shown neuroprotective effects in preclinical models and is used for mechanistic studies and pharmacology experiments. The compound is supplied as a powder for laboratory research and requires appropriate solvent formulation for biological testing.
  • Redox-active oxidoreductase inhibitor.
  • Demonstrated neuroprotective activity in preclinical models.
  • Suitable for in vitro and in vivo pharmacology studies.
  • High reported purity (approximately 99.3%).
  • Soluble in DMSO; established in vivo formulation options reported.
  • Powder stable under low-temperature storage conditions.