Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (2R)-2-hydroxy-2-methyl-4-(2,4,5-trimethyl-3,6-dioxocyclohexa-1,4-dien-1-yl)butanamide | 1147883-03-1 | MFCD00824427 | 99.3% | 265.30 g/mol | C14H19NO4 | 100MG
SDP

Catalog No. 5000236544
Encompass_Preferred

EPI-589 is a redox-active, quinone-derived neuroprotective small molecule (CAS 1147883-03-1) used in preclinical research to study neurodegenerative disorders. It functions as an oxidoreductase inhibitor and free-radical scavenger, demonstrates blood-brain barrier penetration and oral activity, and is supplied as a high-purity research reagent.

  • High purity suitable for research (99.34%).
  • Molecular formula C14H19NO4.
  • Molecular weight 265.30 g/mol.
  • Soluble in DMSO.
  • Available in small research quantities, including 100 mg.
  • Used in neuroprotection and motor neuron disease models.

Catalog No. 50-002-36544 Supplier Medchemexpress LLC Supplier No. HY125999100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

EPI-589 is a redox-active, quinone-derived neuroprotective small molecule (CAS 1147883-03-1) used in preclinical research to study neurodegenerative disorders. It functions as an oxidoreductase inhibitor and free-radical scavenger, demonstrates blood-brain barrier penetration and oral activity, and is supplied as a high-purity research reagent.

  • High purity suitable for research (99.34%).
  • Molecular formula C14H19NO4.
  • Molecular weight 265.30 g/mol.
  • Soluble in DMSO.
  • Available in small research quantities, including 100 mg.
  • Used in neuroprotection and motor neuron disease models.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.