Learn More
Medchemexpress LLC [(2R)-2-hydroxy-3-tetracosanoyloxypropyl] 2-(trimethylazaniumyl)ethyl phosphate | 325171-59-3 | MFCD00674360 | 99.7% | 607.84 g/mol | C32H66NO7P | 5 MG
SDP
Catalog No. 5000222171
Encompass_Preferred
24:0 Lyso PC is a lysophospholipid supplied as a purified white to off-white solid for research use. It is used in studies of lipid metabolism and to formulate lipid nanoparticles for nucleic acid (mRNA) delivery, and is provided with documented chemical identity and stability data.
  • Lysophospholipid useful for lipid nanoparticle formulation and mRNA delivery studies.
  • High purity suitable for analytical and formulation research.
  • White to off-white solid form, convenient for handling and storage.
  • Stable under recommended storage conditions (powder at -20°C up to 3 years).
  • Defined chemical identity and molecular weight for reproducible results.
Catalog No. 50-002-22171 Supplier Medchemexpress LLC Supplier No. HY1386225MG
24:0 Lyso PC is a lysophospholipid supplied as a purified white to off-white solid for research use. It is used in studies of lipid metabolism and to formulate lipid nanoparticles for nucleic acid (mRNA) delivery, and is provided with documented chemical identity and stability data.
  • Lysophospholipid useful for lipid nanoparticle formulation and mRNA delivery studies.
  • High purity suitable for analytical and formulation research.
  • White to off-white solid form, convenient for handling and storage.
  • Stable under recommended storage conditions (powder at -20°C up to 3 years).
  • Defined chemical identity and molecular weight for reproducible results.