Learn More
Strem, An Ascensus Company CAS# 1361315-26-5. 500mg. (2R,2'R)-(-)-[N,N'-Bis(2-pyridylmethyl]-2,2'-bipyrrolidinebis(acetonitrile)iron(II) hexafluoroantimonate Fe(R,R-PDP) White-Chen Catalyst. MFCD16038114
SDP
Supplier:  Strem, An Ascensus Company 260061500MG
Encompass
CAS# 1361315-26-5. 500mg. (2R,2'R)-(-)-[N,N'-Bis(2-pyridylmethyl]-2,2'-bipyrrolidinebis(acetonitrile)iron(II) hexafluoroantimonate Fe(R,R-PDP) White-Chen Catalyst. MFCD16038114. Molecular Weight: 931.90. Molecular Formula: [C24H32FeN6](SbF6)2. Color/form: purple pwdr. Strem# 26-0061. http://www.strem.com/catalog/v/26-0061/
Catalog No. NC1706769