Learn More
Medchemexpress LLC (2R)-3-(1H-indol-3-yl)-2-[[5-[2-(4-methylphenyl)ethynyl]thiophen-2-yl]sulfonylamino]propanoic acid | 203640-27-1 | MFCD22200275 | 99.8% | 464.55 g/mol | C24H20N2O4S2 | 5 MG
SDP
Catalog No. 5000240543
Encompass_Preferred
S 3304 is a research-grade small-molecule inhibitor selective for matrix metalloproteinases MMP-2 and MMP-9. It is provided for in vitro laboratory use in solid form or as a 10 mM solution in DMSO, and is accompanied by analytical data for verification. The compound is intended for studies of extracellular matrix remodeling, invasion, and related signaling pathways.
  • High purity (99.8%) suitable for research assays.
  • Selective inhibition of MMP-2 and MMP-9.
  • Available as solid packs and 10 mM solution in DMSO.
  • Characterized by LC-MS and H NMR data.
  • Supplied with data sheet, certificate of analysis, and safety data sheets.
  • Suitable for in vitro studies of matrix remodeling and tumor invasion.
Catalog No. 50-002-40543 Supplier Medchemexpress LLC Supplier No. HY1069925MG
S 3304 is a research-grade small-molecule inhibitor selective for matrix metalloproteinases MMP-2 and MMP-9. It is provided for in vitro laboratory use in solid form or as a 10 mM solution in DMSO, and is accompanied by analytical data for verification. The compound is intended for studies of extracellular matrix remodeling, invasion, and related signaling pathways.
  • High purity (99.8%) suitable for research assays.
  • Selective inhibition of MMP-2 and MMP-9.
  • Available as solid packs and 10 mM solution in DMSO.
  • Characterized by LC-MS and H NMR data.
  • Supplied with data sheet, certificate of analysis, and safety data sheets.
  • Suitable for in vitro studies of matrix remodeling and tumor invasion.