Learn More
Medchemexpress LLC (2R)-3-(1H-indol-3-yl)-2-[[5-[2-(4-methylphenyl)ethynyl]thiophen-2-yl]sulfonylamino]propanoic acid | 203640-27-1 | MFCD22200275 | 99.8% | 464.56 g/mol | C24H20N2O4S2 | 25 MG
SDP
Catalog No. 5000240589
Encompass_Preferred
S 3304 is a matrix metalloproteinase inhibitor selective for MMP-2 and MMP-9 that contains an alkyne functional group enabling copper-catalyzed azide-alkyne cycloaddition (CuAAC) for bioorthogonal labeling. It is supplied as a light yellow solid with high purity for research use.
  • Selective inhibitor of MMP-2 and MMP-9
  • Contains an alkyne for copper-catalyzed azide-alkyne cycloaddition
  • High purity (99.8%) suitable for biochemical studies
  • Light yellow solid form for convenient handling
  • Stable under recommended storage conditions
  • Available in small research pack sizes and solution form
Catalog No. 50-002-40589 Supplier Medchemexpress LLC Supplier No. HY10699225MG
S 3304 is a matrix metalloproteinase inhibitor selective for MMP-2 and MMP-9 that contains an alkyne functional group enabling copper-catalyzed azide-alkyne cycloaddition (CuAAC) for bioorthogonal labeling. It is supplied as a light yellow solid with high purity for research use.
  • Selective inhibitor of MMP-2 and MMP-9
  • Contains an alkyne for copper-catalyzed azide-alkyne cycloaddition
  • High purity (99.8%) suitable for biochemical studies
  • Light yellow solid form for convenient handling
  • Stable under recommended storage conditions
  • Available in small research pack sizes and solution form