Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2R,3R)-1-Carboxy-4-iodo-2,3-dihydroxycyclohexa-4,6-diene, 95%, Thermo Scientific™

Catalog No. AC271622500
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC271622500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC271622500 Supplier Thermo Scientific Chemicals Supplier No. 271622500

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 205503-96-4
Molecular Formula C7H7IO4
Molecular Weight (g/mol) 282.033
MDL Number MFCD00142968
InChI Key OJVUEUHUFZSXFM-RITPCOANSA-N
Synonym 2r,3r-1-carboxy-4-iodo-2,3-dihydroxycyclohexa-4,6-diene, 5r,6r-5,6-dihydroxy-4-iodocyclohexa-1,3-diene-1-carboxylic acid, 5r,6r-5,6-dihydroxy-4-iodo-1,3-cyclohexadiene-1-carboxylic acid, 5r,6r-5,6-dihydroxy-4-iodocyclohexa-1,3-dienecarboxylic acid, 2r,3r-1-carboxy-4-iodo-2,3-dihydroxycyclohexa-4,6-diene, 95
PubChem CID 2724721
IUPAC Name (5R,6R)-5,6-dihydroxy-4-iodocyclohexa-1,3-diene-1-carboxylic acid
SMILES C1=C(C(C(C(=C1)I)O)O)C(=O)O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 95%
Quantity 250 mg
Formula Weight 282.02
Percent Purity ≥94%
Physical Form Powder
Chemical Name or Material (2R,3R)-1-Carboxy-4-iodo-2,3-dihydroxycyclohexa-4,6-diene, 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.