Learn More
Medchemexpress LLC (2R,3R,3aS,7aR,9R,10R,10aS,14aR)-2,9-Bis(6-amino-9H-purin-9-yl)-3,5,10,12-tetrahydroxyoctahydro | 99.4% | 692.47 | 25 MG
SDP
Catalog No. 5000438325
Encompass_Preferred
c-di-AMP diammonium is a STING agonist that binds to the transmembrane protein STING, activating the TBK3-IRF3 signaling pathway and subsequently triggering the production of type I IFN and TNF. It also functions as a bacterial second messenger, regulating cell growth, survival, and virulence primarily in Gram-positive bacteria, and modulating the host immune response. Furthermore, c-di-AMP diammonium acts as a potent mucosal adjuvant, stimulating both humoral and cellular immune responses.
  • STING agonist, activating the TBK3-IRF3 signaling pathway.
  • Triggers the production of type I IFN and TNF.
  • Functions as a bacterial second messenger, regulating cell growth, survival, and virulence in Gram-positive bacteria.
  • Regulates host immune response.
  • Potent mucosal adjuvant, stimulating both humoral and cellular responses.
Catalog No. 50-004-38325 Supplier Medchemexpress LLC Supplier No. 25MGHY12326B
c-di-AMP diammonium is a STING agonist that binds to the transmembrane protein STING, activating the TBK3-IRF3 signaling pathway and subsequently triggering the production of type I IFN and TNF. It also functions as a bacterial second messenger, regulating cell growth, survival, and virulence primarily in Gram-positive bacteria, and modulating the host immune response. Furthermore, c-di-AMP diammonium acts as a potent mucosal adjuvant, stimulating both humoral and cellular immune responses.
  • STING agonist, activating the TBK3-IRF3 signaling pathway.
  • Triggers the production of type I IFN and TNF.
  • Functions as a bacterial second messenger, regulating cell growth, survival, and virulence in Gram-positive bacteria.
  • Regulates host immune response.
  • Potent mucosal adjuvant, stimulating both humoral and cellular responses.